siRNA Details
| Gene Symbol: ALDH3B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000694.3 | NP_000685.1 | 10 | 2331 | ||
| NM_001030010.2 | NP_001025181.1 | 9 | 2220 | |||
| NM_001161473.2 | NP_001154945.1 | 10 | 2389 | |||
| NM_001290058.1 | NP_001276987.1 | 9 | 1980 | |||
| NM_001290059.1 | NP_001276988.1 | 9 | 2249 | |||
| GenBank mRNA | AB209651.1 | - | 8 | 4486 | ||
| BC013584.2 | - | 10 | 2284 | 3878124 | ||
| BC014168.2 | - | 4 | 1319 | |||
| U10868.1 | - | 10 | 2292 | |||

ALDH3B1
Genomic Map