siRNA Details
| Gene Symbol: CHGB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001819.2 | NP_001810.2 | 2, 3 | 405 | ||
| GenBank mRNA | AB209712.1 | - | 2, 3 | 202 | ||
| AK054871.1 | - | 1, 2 | 878 | |||
| AK225125.1 | - | 2, 3 | 223 | |||
| AK289386.1 | - | 2, 3 | 225 | |||
| AK293536.1 | - | 2, 3 | 223 | |||
| AK312237.1 | - | 2, 3 | 225 | |||
| BC000375.2 | - | 2, 3 | 203 | 2820258 | ||
| CR456726.1 | - | 2, 3 | 99 | |||
| DC312240.1 | - | 2, 3 | 405 | |||
| Y00064.1 | - | 2, 3 | 211 | |||

CHGB
Genomic Map