siRNA Details
| Gene Symbol: SLC2A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042.2 | NP_001033.1 | 11 | 2085 | ||
| GenBank mRNA | BC034387.1 | - | 11 | 2146 | 5187454 | |
| M20747.1 | - | 11 | 2030 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC2A4| Gene Symbol: SLC2A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042.2 | NP_001033.1 | 11 | 2085 | ||
| GenBank mRNA | BC034387.1 | - | 11 | 2146 | 5187454 | |
| M20747.1 | - | 11 | 2030 | |||