siRNA Details
| Gene Symbol: COL19A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001858.5 | NP_001849.2 | 3, 4 | 291 | ||
| XM_011535437.2 | XP_011533739.1 | 3, 4 | 558 | |||
| XM_011535438.2 | XP_011533740.1 | 3, 4 | 286 | |||
| XM_017010253.1 | XP_016865742.1 | 3, 4 | 558 | |||
| XM_017010254.1 | XP_016865743.1 | 3, 4 | 558 | |||
| XM_017010255.1 | XP_016865744.1 | 3, 4 | 497 | |||
| XM_017010256.1 | XP_016865745.1 | 3, 4 | 291 | |||
| XM_017010257.1 | XP_016865746.1 | 3, 4 | 558 | |||
| XM_017010258.1 | XP_016865747.1 | 3, 4 | 291 | |||
| XM_017010259.1 | XP_016865748.1 | 3, 4 | 557 | |||
| GenBank mRNA | AK310621.1 | - | 3, 4 | 281 | ||
| BC113362.1 | - | 3, 4 | 259 | 8322414 | ||
| BC113364.1 | - | 3, 4 | 259 | 8322416 | ||
| D38163.1 | - | 3, 4 | 291 | |||
| M63597.1 | - | 2, 3 | 100 | |||

COL19A1
Genomic Map