siRNA Details
| Gene Symbol: GRID2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286838.1 | NP_001273767.1 | 15 | 2749 | ||
| NM_001510.3 | NP_001501.2 | 16 | 3034 | |||
| XM_011531893.2 | XP_011530195.2 | 16 | 9271 | |||
| XM_011531894.2 | XP_011530196.1 | 14 | 2310 | |||
| XM_011531895.2 | XP_011530197.1 | 14 | 2227 | |||
| XM_017008118.1 | XP_016863607.1 | 15 | 9223 | |||
| XM_017008119.1 | XP_016863608.1 | 16 | 2808 | |||
| XM_017008123.1 | XP_016863612.1 | 14 | 3538 | |||
| XM_017008124.1 | XP_016863613.1 | 13 | 2758 | |||
| XM_017008125.1 | XP_016863614.1 | 13 | 2255 | |||
| XM_017008126.1 | XP_016863615.1 | 12 | 1971 | |||
| GenBank mRNA | AB209318.1 | - | 16 | 3034 | ||
| AF009014.1 | - | 16 | 2776 | |||
| BC099652.3 | - | 16 | 2776 | 40008396 | ||
| BC099653.3 | - | 16 | 2776 | 40008397 | ||
| BC099654.3 | - | 15 | 2491 | 40008398 | ||

GRID2
Genomic Map