siRNA Details
| Gene Symbol: GTF2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001514.5 | NP_001505.1 | 7 | 1197 | ||
| XM_011541299.1 | XP_011539601.1 | 7 | 1399 | |||
| GenBank mRNA | AK289822.1 | - | 7 | 1143 | ||
| BC020597.1 | - | 7 | 1143 | 4807599 | ||
| M76766.1 | - | 7 | 1116 | |||
| S44184.1 | - | 6 | 1066 | |||

GTF2B
Genomic Map