siRNA Details
| Gene Symbol: ID4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001546.3 | NP_001537.1 | 3 | 1915 | ||
| GenBank mRNA | AJ420553.1 | - | 1 | 169 | ||
| AK130851.1 | - | 1 | 288 | |||
| BC014941.1 | - | 3 | 1899 | 4552357 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ID4| Gene Symbol: ID4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001546.3 | NP_001537.1 | 3 | 1915 | ||
| GenBank mRNA | AJ420553.1 | - | 1 | 169 | ||
| AK130851.1 | - | 1 | 288 | |||
| BC014941.1 | - | 3 | 1899 | 4552357 | ||