siRNA Details
| Gene Symbol: CD69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001781.2 | NP_001772.1 | 5 | 864 | ||
| GenBank mRNA | AK291869.1 | - | 5 | 864 | ||
| BC007037.1 | - | 5 | 864 | 3950403 | ||
| Z22576.1 | - | 5 | 864 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CD69| Gene Symbol: CD69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001781.2 | NP_001772.1 | 5 | 864 | ||
| GenBank mRNA | AK291869.1 | - | 5 | 864 | ||
| BC007037.1 | - | 5 | 864 | 3950403 | ||
| Z22576.1 | - | 5 | 864 | |||