siRNA Details
| Gene Symbol: FNTB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002028.3 | NP_002019.1 | 12 | 1394 | ||
| GenBank mRNA | AK093298.1 | - | 10 | 1152 | ||
| AK225917.1 | - | 12 | 1261 | |||
| AK295972.1 | - | 9 | 958 | |||
| AK315714.1 | - | 12 | 1461 | |||
| BC020232.1 | - | 12 | 1244 | 4652202 | ||
| BX248269.1 | - | 12 | 1240 | |||
| L00635.1 | - | 12 | 1248 | |||
| L10414.1 | - | 11 | 1079 | |||
| Gene Symbol: CHURC1-FNTB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001202558.1 | NP_001189487.1 | 13 | 1395 | ||
| NM_001202559.1 | NP_001189488.1 | 14 | 1466 | |||
| GenBank mRNA | AK296850.1 | - | 14 | 1334 | ||
| AK303739.1 | - | 13 | 1271 | |||

FNTB
Genomic Map