siRNA Details
Gene Symbol: IRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002199.3 | NP_002190.2 | 9 | 1485 | ||
GenBank mRNA | BC015803.2 | - | 8 | 1292 | 3920890 | |
X15949.1 | - | 9 | 1384 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: IRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002199.3 | NP_002190.2 | 9 | 1485 | ||
GenBank mRNA | BC015803.2 | - | 8 | 1292 | 3920890 | |
X15949.1 | - | 9 | 1384 |