siRNA Details
Gene Symbol: CTSE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317331.1 | NP_001304260.1 | 4 | 555 | ||
NM_001910.3 | NP_001901.1 | 5 | 712 | |||
NM_148964.2 | NP_683865.1 | 5 | 712 | |||
XM_011509244.1 | XP_011507546.1 | 5 | 717 | |||
XM_011509245.1 | XP_011507547.1 | 5 | 717 | |||
GenBank mRNA | AJ250716.1 | - | 5 | 698 | ||
AJ250717.1 | - | 5 | 698 | |||
AK292057.1 | - | 5 | 642 | |||
AK298069.1 | - | 4 | 408 | |||
BC042537.1 | - | 5 | 679 | 5174814 | ||
J05036.1 | - | 5 | 643 |
