siRNA Details
| Gene Symbol: MRC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002438.3 | NP_002429.1 | 7 | 1314 | ||
| GenBank mRNA | J05550.1 | - | 7 | 1294 | ||
| X55635.1 | - | 7 | 1267 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MRC1| Gene Symbol: MRC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002438.3 | NP_002429.1 | 7 | 1314 | ||
| GenBank mRNA | J05550.1 | - | 7 | 1294 | ||
| X55635.1 | - | 7 | 1267 | |||