siRNA Details
| Gene Symbol: DAO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001917.4 | NP_001908.3 | 5 | 581 | ||
| XM_011538004.2 | XP_011536306.1 | 6 | 667 | |||
| XM_011538005.2 | XP_011536307.1 | 6 | 671 | |||
| GenBank mRNA | AK312995.1 | - | 5 | 568 | ||
| AX741655.1 | - | 5 | 571 | |||
| AX741656.1 | - | 5 | 571 | |||
| AX741658.1 | - | 4 | 456 | |||
| AX776478.1 | - | 5 | 571 | |||
| AX776479.1 | - | 5 | 571 | |||
| AX776480.1 | - | 8 | 1583 | |||
| AX786386.1 | - | 5 | 571 | |||
| BC029057.1 | - | 5 | 551 | 5184628 | ||
| BC074770.2 | - | 5 | 453 | 30915193 | ||
| BX538210.1 | - | 5 | 513 | |||
| KU178015.1 | - | 4 | 428 | |||
| KU178016.1 | - | 4 | 428 | |||

DAO
Genomic Map