siRNA Details
| Gene Symbol: NDUFA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002490.3 | NP_002481.2 | 3 | 941 | ||
| GenBank mRNA | BC002772.1 | - | 3 | 802 | 3619356 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

NDUFA6| Gene Symbol: NDUFA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002490.3 | NP_002481.2 | 3 | 941 | ||
| GenBank mRNA | BC002772.1 | - | 3 | 802 | 3619356 | |