siRNA Details
| Gene Symbol: NDUFB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199957.1 | NP_001186886.1 | 4 | 371 | ||
| NM_001199958.1 | NP_001186887.1 | 5 | 420 | |||
| NM_002492.3 | NP_002483.1 | 6 | 527 | |||
| GenBank mRNA | AF047181.1 | - | 6 | 504 | ||
| AK313822.1 | - | 6 | 505 | |||
| AW955118.1 | - | 5 | 377 | |||
| BC005271.1 | - | 6 | 518 | 3997377 | ||
| BC009796.1 | - | 6 | 522 | 4284776 | ||
| BC017909.1 | - | 6 | 517 | |||
| BC093071.1 | - | 4 | 348 | 30325282 | ||
| BG776403.1 | - | 6 | 534 | |||
| BX648034.1 | - | 4 | 365 | |||
| DA112751.1 | - | 6 | 530 | |||

NDUFB5
Genomic Map