siRNA Details
| Gene Symbol: OLR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172632.1 | NP_001166103.1 | 5 | 1817 | ||
| NM_001172633.1 | NP_001166104.1 | 5 | 1841 | |||
| NM_002543.3 | NP_002534.1 | 6 | 1957 | |||
| GenBank mRNA | BC022295.1 | - | 6 | 1896 | 4722086 | |

OLR1
Genomic Map