siRNA Details
| Gene Symbol: PXN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | U14588.1 | - | 11 | 3001 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PXN| Gene Symbol: PXN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | U14588.1 | - | 11 | 3001 | ||