siRNA Details
| Gene Symbol: SLC6A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134367.3 | NP_001127839.2 | 15 | 2689 | ||
| NM_003043.5 | NP_003034.2 | 15 | 2693 | |||
| NR_103507.2 | 14 | 2568 | ||||
| XM_006713307.2 | XP_006713370.1 | 14 | 2497 | |||
| XM_011534030.2 | XP_011532332.1 | 14 | 2450 | |||
| XM_011534031.2 | XP_011532333.1 | 16 | 2821 | |||
| GenBank mRNA | AB209172.1 | - | 14 | 2493 | ||
| BC137128.1 | - | 15 | 2642 | 9021125 | ||
| BC137129.1 | - | 15 | 2640 | 9021126 | ||
| BX509263.1 | - | 1 | 512 | |||
| U16120.1 | - | 16 | 2742 | |||
| Z18956.1 | - | 13 | 2388 | |||

SLC6A6
Genomic Map