siRNA Details
| Gene Symbol: SLIT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271946.1 | NP_001258875.1 | 18 | 2266 | ||
| NM_003062.3 | NP_003053.1 | 18 | 2266 | |||
| XM_017009779.1 | XP_016865268.1 | 18 | 1669 | |||
| GenBank mRNA | AB011538.2 | - | 18 | 2266 | ||
| AB017169.1 | - | 18 | 2109 | |||
| AF075240.1 | - | 2 | 127 | |||
| AL122074.1 | - | 8 | 3550 | |||
| BC146759.1 | - | 18 | 2266 | 9007197 | ||

SLIT3
Genomic Map