siRNA Details
| Gene Symbol: TRIM21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003141.3 | NP_003132.2 | 6 | 901 | ||
| GenBank mRNA | AK225564.1 | - | 4 | 614 | ||
| BC010861.1 | - | 6 | 872 | 3900540 | ||
| M34551.1 | - | 5 | 826 | |||
| M62800.1 | - | 6 | 873 | |||

TRIM21
Genomic Map