siRNA Details
| Gene Symbol: STAT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243835.1 | NP_001230764.1 | 20 | 2036 | ||
| NM_003151.3 | NP_003142.1 | 20 | 2087 | |||
| XM_006712719.3 | XP_006712782.1 | 20 | 2082 | |||
| XM_011511705.2 | XP_011510007.1 | 23 | 2233 | |||
| GenBank mRNA | AB209043.1 | - | 10 | 723 | ||
| AK299976.1 | - | 17 | 1568 | |||
| AK300868.1 | - | 20 | 1840 | |||
| AK301172.1 | - | 18 | 1511 | |||
| BC031212.1 | - | 20 | 2008 | 4830583 | ||
| L78440.1 | - | 20 | 1853 | |||

STAT4
Genomic Map