siRNA Details
| Gene Symbol: TFRC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128148.2 | NP_001121620.1 | 16, 17 | 1827 | ||
| NM_001313965.1 | NP_001300894.1 | 15, 16 | 1625 | |||
| NM_001313966.1 | NP_001300895.1 | 14, 15 | 1429 | |||
| NM_003234.3 | NP_003225.2 | 16, 17 | 1968 | |||
| GenBank mRNA | AB209254.1 | - | 16, 17 | 1731 | ||
| AK291723.1 | - | 16, 17 | 1968 | |||
| AK294758.1 | - | 14, 15 | 1359 | |||
| BC001188.1 | - | 16, 17 | 1741 | 3354176 | ||
| BX537966.1 | - | 15, 16 | 1555 | |||
| M11507.1 | - | 16, 17 | 1780 | |||
| X01060.1 | - | 16, 17 | 1948 | |||

TFRC
Genomic Map