siRNA Details
| Gene Symbol: VLDLR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001018056.2 | NP_001018066.1 | 18 | 2989 | ||
| NM_001322225.1 | NP_001309154.1 | 18 | 2950 | |||
| NM_001322226.1 | NP_001309155.1 | 17 | 2866 | |||
| NM_003383.4 | NP_003374.3 | 19 | 3073 | |||
| GenBank mRNA | AB208822.1 | - | 19 | 2680 | ||
| AK092381.1 | - | 17 | 2340 | |||
| AK095744.1 | - | 14 | 2069 | |||
| AK289527.1 | - | 10 | 1372 | |||
| BC136562.1 | - | 19 | 2794 | 9020552 | ||
| BC142653.1 | - | 18 | 2651 | 40148037 | ||
| BC144245.1 | - | 18 | 2710 | 9052765 | ||
| D16493.1 | - | 19 | 2727 | |||
| D16494.1 | - | 18 | 2574 | |||
| L20470.1 | - | 19 | 3048 | |||
| L22431.1 | - | 19 | 2770 | |||
| S73849.1 | - | 19 | 2742 | |||

VLDLR
Genomic Map