siRNA Details
| Gene Symbol: ZNF267 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001265588.1 | NP_001252517.1 | 5 | 3299 | ||
| NM_003414.5 | NP_003405.3 | 4 | 3148 | |||
| NR_049749.1 | 5 | 3192 | ||||
| XM_017022865.1 | XP_016878354.1 | 5 | 3235 | |||
| XM_017022866.1 | XP_016878355.1 | 6 | 3219 | |||
| GenBank mRNA | AA570774.1 | - | 1 | 315 | ||
| AK096390.1 | - | 5 | 1911 | |||
| BC036367.1 | - | 4 | 3072 | 4828269 | ||

ZNF267
Genomic Map