siRNA Details
| Gene Symbol: CHN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025201.3 | NP_001020372.2 | 13 | 1499 | ||
| NM_001206602.1 | NP_001193531.1 | 7 | 1347 | |||
| NM_001822.5 | NP_001813.1 | 13 | 1576 | |||
| NR_038133.1 | 12 | 1101 | ||||
| GenBank mRNA | AB209788.1 | - | 12 | 1086 | ||
| AK055060.1 | - | 7 | 1278 | |||
| AK289941.1 | - | 13 | 1506 | |||
| AK300890.1 | - | 13 | 1493 | |||
| BC011393.1 | - | 13 | 1548 | 3871858 | ||
| CR456864.1 | - | 7 | 755 | |||
| X51408.1 | - | 1307 | ||||
| Z22641.1 | - | 13 | 1245 | |||

CHN1
Genomic Map