siRNA Details
| Gene Symbol: ZNF134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003435.4 | NP_003426.3 | 3 | 1724 | ||
| GenBank mRNA | AK291763.1 | - | 3 | 1693 | ||
| BC011646.2 | - | 2 | 1583 | |||
| BC021280.2 | - | 2 | 1568 | |||
| BC042636.1 | - | 3 | 1683 | 4826076 | ||
| BC112294.1 | - | 3 | 1484 | 8327762 | ||
| BC113410.1 | - | 3 | 1481 | 8322462 | ||
| U09412.1 | - | 3 | 1666 | |||

ZNF134
Genomic Map