siRNA Details
| Gene Symbol: ZNF155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001260486.1 | NP_001247415.1 | 5 | 2225 | ||
| NM_001260487.1 | NP_001247416.1 | 5 | 2108 | |||
| NM_001260488.1 | NP_001247417.1 | 6 | 2211 | |||
| NM_003445.3 | NP_003436.3 | 5 | 2199 | |||
| NM_198089.2 | NP_932355.2 | 5 | 2122 | |||
| XM_005259215.3 | XP_005259272.1 | 5 | 2269 | |||
| XM_011527278.2 | XP_011525580.1 | 6 | 2154 | |||
| XM_011527279.2 | XP_011525581.1 | 6 | 3214 | |||
| XM_017027248.1 | XP_016882737.1 | 6 | 2246 | |||
| GenBank mRNA | AK022875.1 | - | 5 | 2089 | ||
| AK297360.1 | - | 6 | 2187 | |||

ZNF155
Genomic Map