siRNA Details
| Gene Symbol: ZNF282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303481.1 | NP_001290410.1 | 9 | 2893 | ||
| NM_003575.3 | NP_003566.1 | 8 | 3150 | |||
| XM_006716151.3 | XP_006716214.1 | 8 | 3147 | |||
| GenBank mRNA | BC031686.1 | - | 4 | 1905 | ||
| BC041704.1 | - | 8 | 2877 | |||
| BC055287.1 | - | 8 | 2942 | |||
| D30612.1 | - | 8 | 3161 | |||

ZNF282
Genomic Map