siRNA Details
| Gene Symbol: COX6B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001863.4 | NP_001854.1 | 4 | 547 | ||
| GenBank mRNA | BC001015.2 | - | 4 | 517 | 3344701 | |
| BC002478.2 | - | 4 | 416 | 3347455 | ||
| X13923.1 | - | 4 | 399 | |||
| X54473.1 | - | 4 | 402 | |||

COX6B1
Genomic Map