siRNA Details
| Gene Symbol: CPM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005502.2 | NP_001005502.1 | 9 | 1221 | ||
| NM_001874.4 | NP_001865.1 | 9 | 1253 | |||
| NM_198320.3 | NP_938079.1 | 9 | 1239 | |||
| GenBank mRNA | AF368463.1 | - | 9 | 1211 | ||
| AK130146.1 | - | 6 | 829 | |||
| AK313180.1 | - | 9 | 1211 | |||
| BC022276.1 | - | 9 | 1211 | 4768771 | ||
| J04970.2 | - | 9 | 1253 | |||

CPM
Genomic Map