siRNA Details
Gene Symbol: EPS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004447.5 | NP_004438.3 | 5 | 736 | ||
XM_011520605.2 | XP_011518907.1 | 6 | 796 | |||
GenBank mRNA | AK291777.1 | - | 5 | 541 | ||
AK292931.1 | - | 5 | 393 | |||
AK304860.1 | - | 5 | 736 | |||
BC030010.2 | - | 5 | 548 | 4820933 | ||
U12535.1 | - | 5 | 508 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
