siRNA Details
| Gene Symbol: EPS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004447.5 | NP_004438.3 | 21 | 3210 | ||
| XM_011520605.2 | XP_011518907.1 | 22 | 3269 | |||
| GenBank mRNA | AK292931.1 | - | 21 | 2867 | ||
| BC030010.2 | - | 21 | 3022 | 4820933 | ||
| U12535.1 | - | 21 | 2982 | |||

EPS8
Genomic Map