siRNA Details
| Gene Symbol: NOTCH4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004557.3 | NP_004548.3 | 8 | 1566 | ||
| NR_134949.1 | 8 | 1566 | ||||
| NR_134950.1 | 8 | 1566 | ||||
| GenBank mRNA | AK290785.1 | - | 8 | 1563 | ||
| BC063815.1 | - | 8 | 1556 | |||
| BC140782.1 | - | 8 | 1521 | 9021650 | ||
| BC144540.1 | - | 8 | 1527 | |||
| BC144541.1 | - | 8 | 1521 | |||
| U95299.1 | - | 8 | 1517 | |||

NOTCH4
Genomic Map