siRNA Details
| Gene Symbol: ZFYVE9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004799.3 | NP_004790.2 | 4 | 1651 | ||
| NM_007324.3 | NP_015563.2 | 4 | 1651 | |||
| XM_011542437.2 | XP_011540739.1 | 4 | 1522 | |||
| XM_017002845.1 | XP_016858334.1 | 4 | 1744 | |||
| GenBank mRNA | AB209125.1 | - | 1 | 925 | ||
| AF104304.1 | - | 3 | 1331 | |||
| AF130419.1 | - | 4 | 1371 | |||
| AF130420.1 | - | 4 | 1371 | |||
| BC032680.1 | - | 2 | 1210 | 5585817 | ||

ZFYVE9
Genomic Map