siRNA Details
Gene Symbol: PGGT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005023.3 | NP_005014.2 | 9 | 1540 | ||
XM_005272020.3 | XP_005272077.2 | 10 | 1443 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PGGT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005023.3 | NP_005014.2 | 9 | 1540 | ||
XM_005272020.3 | XP_005272077.2 | 10 | 1443 |