siRNA Details
| Gene Symbol: DCC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005215.3 | NP_005206.2 | 2 | 978 | ||
| XM_011525843.1 | XP_011524145.1 | 2 | 978 | |||
| XM_017025568.1 | XP_016881057.1 | 2 | 978 | |||
| XM_017025569.1 | XP_016881058.1 | 2 | 978 | |||
| GenBank mRNA | AK302643.1 | - | 1 | 158 | ||
| BC036524.1 | - | 1 | 164 | |||
| M32292.1 | - | 2 | 368 | |||
| X76132.1 | - | 2 | 362 | |||

DCC
Genomic Map