siRNA Details
| Gene Symbol: TP53BP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031685.2 | NP_001026855.2 | 8 | 1181 | ||
| NM_005426.2 | NP_005417.1 | 9 | 1313 | |||
| XM_011544268.1 | XP_011542570.1 | 7 | 1042 | |||
| XM_011544269.2 | XP_011542571.1 | 5 | 700 | |||
| XM_017002220.1 | XP_016857709.1 | 9 | 1322 | |||
| GenBank mRNA | AJ318888.2 | - | 8 | 1126 | ||
| AK294432.1 | - | 8 | 1033 | |||
| BC040247.1 | - | 6 | 657 | 4454266 | ||
| BC046150.1 | - | 8 | 1124 | |||
| BC058918.1 | - | 8 | 1139 | 4537206 | ||
| U58334.1 | - | 9 | 1258 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_173378, BC042874, CN529251, AK134556.

TP53BP2
Genomic Map