siRNA Details
| Gene Symbol: TLX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195517.1 | NP_001182446.1 | 3 | 1765 | ||
| NM_005521.3 | NP_005512.1 | 3 | 1742 | |||
| XM_011539744.2 | XP_011538046.1 | 3 | 2294 | |||
| GenBank mRNA | BC130530.1 | - | 3 | 1765 | 40146561 | |
| M62626.1 | - | 3 | 2522 | |||
| M75952.1 | - | 3 | 1514 | |||
| S38742.1 | - | 3 | 1602 | |||

TLX1
Genomic Map