siRNA Details
| Gene Symbol: TCF20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005650.3 | NP_005641.1 | 2 | 1051 | ||
| NM_181492.2 | NP_852469.1 | 1 | 950 | |||
| XM_005261722.3 | XP_005261779.1 | 2 | 2254 | |||
| XM_006724313.3 | XP_006724376.1 | 2 | 1235 | |||
| XM_011530353.2 | XP_011528655.1 | 2 | 1235 | |||
| XM_011530354.2 | XP_011528656.1 | 2 | 1235 | |||
| GenBank mRNA | AB006630.1 | - | 1 | 183 | ||
| KF851355.1 | - | 2 | 1051 | |||

TCF20
Genomic Map