siRNA Details
| Gene Symbol: ZNF263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005741.4 | NP_005732.2 | 6 | 2544 | ||
| XM_006720831.1 | XP_006720894.1 | 6 | 2612 | |||
| XM_006720832.1 | XP_006720895.1 | 5 | 2534 | |||
| XM_006720833.1 | XP_006720896.1 | 4 | 2396 | |||
| XM_006720834.3 | XP_006720897.1 | 5 | 2667 | |||
| GenBank mRNA | AK295350.1 | - | 2 | 1410 | ||
| BC008805.2 | - | 6 | 2542 | 3945548 | ||
| D88827.1 | - | 6 | 2504 | |||

ZNF263
Genomic Map