siRNA Details
| Gene Symbol: DSC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001941.4 | NP_001932.2 | 2 | 265 | ||
| NM_024423.3 | NP_077741.2 | 2 | 265 | |||
| GenBank mRNA | AK291748.1 | - | 2 | 191 | ||
| D17427.1 | - | 2 | 177 | |||
| DA860471.1 | - | 2 | 265 | |||
| X83929.1 | - | 3 | 131 | |||

DSC3
Genomic Map