siRNA Details
| Gene Symbol: AASS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005763.3 | NP_005754.2 | 5 | 659 | ||
| XM_011515725.2 | XP_011514027.1 | 5 | 638 | |||
| GenBank mRNA | AF229180.1 | - | 5 | 583 | ||
| AJ007714.2 | - | 4 | 610 | |||
| BC144247.1 | - | 4 | 569 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_013930, BC005420, AK098141, AK044323, AJ224761, and others.

AASS
Genomic Map