siRNA Details
| Gene Symbol: ZNF256 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005773.2 | NP_005764.2 | 3 | 2121 | ||
| XM_017026138.1 | XP_016881627.1 | 2 | 2018 | |||
| GenBank mRNA | AF067165.1 | - | 2 | 1830 | ||
| BC001438.1 | - | 3 | 2083 | 3138611 | ||

ZNF256
Genomic Map