siRNA Details
| Gene Symbol: PDGFRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006207.2 | NP_006198.1 | 3 | 745 | ||
| GenBank mRNA | AK289450.1 | - | 3 | 745 | ||
| BC010927.1 | - | 2 | 316 | 4278882 | ||
| CR541952.1 | - | 2 | 300 | |||
| D37965.1 | - | 2 | 361 | |||

PDGFRL
Genomic Map