siRNA Details
| Gene Symbol: PDGFRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006207.2 | NP_006198.1 | 6, 7 | 1392 | ||
| GenBank mRNA | AK289450.1 | - | 6, 7 | 1392 | ||
| BC010927.1 | - | 5, 6 | 963 | 4278882 | ||
| CR541952.1 | - | 5, 6 | 947 | |||
| D37965.1 | - | 5, 6 | 1008 | |||

PDGFRL
Genomic Map