siRNA Details
| Gene Symbol: ENO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193503.1 | NP_001180432.1 | 3 | 239 | ||
| NM_001976.4 | NP_001967.3 | 3 | 281 | |||
| NM_053013.3 | NP_443739.3 | 3 | 239 | |||
| XM_005256521.2 | XP_005256578.1 | 3 | 667 | |||
| XM_011523729.1 | XP_011522031.1 | 3 | 257 | |||
| XM_017024346.1 | XP_016879835.1 | 3 | 674 | |||
| GenBank mRNA | AK300662.1 | - | 3 | 209 | ||
| AK300673.1 | - | 3 | 251 | |||
| AK300709.1 | - | 3 | 209 | |||
| AK309864.1 | - | 4 | 332 | |||
| AU143784.1 | - | 3 | 278 | |||
| BC017249.2 | - | 3 | 196 | 4902149 | ||
| BJ995304.1 | - | 3 | 255 | |||
| DA082822.1 | - | 3 | 239 | |||
| DA566866.1 | - | 3 | 237 | |||
| X16504.1 | - | 2 | 143 | |||
| X51957.1 | - | 2 | 155 | |||

ENO3
Genomic Map