siRNA Details
| Gene Symbol: ETFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014763.1 | NP_001014763.1 | 1 | 644 | ||
| NM_001985.2 | NP_001976.1 | 2 | 259 | |||
| GenBank mRNA | AF370381.1 | - | 1 | 626 | ||
| AK055285.1 | - | 1 | 646 | |||
| AK291881.1 | - | 2 | 186 | |||
| AL833205.1 | - | 1 | 2920 | |||
| BC093961.1 | - | 2 | 190 | 7939806 | ||
| BC093963.1 | - | 2 | 189 | 7939808 | ||
| BG281641.1 | - | 1 | 77 | |||
| BG399783.1 | - | 2 | 260 | |||
| CR456827.1 | - | 2 | 167 | |||
| X71129.1 | - | 2 | 194 | |||

ETFB
Genomic Map