siRNA Details
| Gene Symbol: EVPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320747.1 | NP_001307676.1 | 3 | 595 | ||
| NM_001988.3 | NP_001979.2 | 3 | 595 | |||
| GenBank mRNA | AK290333.1 | - | 3 | 595 | ||
| BC126103.1 | - | 3 | 374 | 8991819 | ||
| BC126105.1 | - | 3 | 374 | 8991821 | ||
| BC143814.1 | - | 3 | 374 | 9052329 | ||
| U53786.1 | - | 3 | 439 | |||

EVPL
Genomic Map