siRNA Details
| Gene Symbol: HOXD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006898.4 | NP_008829.3 | 3 | 1773 | ||
| XM_005246509.3 | XP_005246566.1 | 4 | 2040 | |||
| XM_005246510.4 | XP_005246567.1 | 4 | 1856 | |||
| XM_005246511.3 | XP_005246568.1 | 4 | 2464 | |||
| XM_005246513.4 | XP_005246570.1 | 4 | 2067 | |||
| XM_006712477.2 | XP_006712540.1 | 3 | 1750 | |||
| XM_011511065.2 | XP_011509367.1 | 4 | 3844 | |||
| XM_011511066.2 | XP_011509368.1 | 4 | 1918 | |||
| GenBank mRNA | BC005124.2 | - | 3 | 1774 | 3936607 | |
| BC008789.2 | - | 2 | 1705 | 3628279 | ||

HOXD3
Genomic Map