siRNA Details
| Gene Symbol: ZNF33B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001305033.1 | NP_001291962.1 | 4 | 5646 | ||
| NM_001305035.1 | NP_001291964.1 | 6 | 5776 | |||
| NM_001305036.1 | NP_001291965.1 | 5 | 5704 | |||
| NM_001305037.1 | NP_001291966.1 | 5 | 5631 | |||
| NM_001305038.1 | NP_001291967.1 | 5 | 5723 | |||
| NM_001305039.1 | NP_001291968.1 | 6 | 5781 | |||
| NM_001305040.1 | NP_001291969.1 | 7 | 5857 | |||
| NM_006955.2 | NP_008886.1 | 5 | 5699 | |||
| GenBank mRNA | AJ491697.1 | - | 5 | 5406 | ||

ZNF33B
Genomic Map